Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGTTAAGAATATTATGACCCA[C/G]AATGTGGAGCGGATCCTGGCCCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607029 MIM: 603177 | ||||||||||||||||||||
Literature Links: |
VAMP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VAMP5 - vesicle associated membrane protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAMP8 - vesicle associated membrane protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003761.4 | 199 | Missense Mutation | CAC,CAG | H,Q 29 | NP_003752.2 | |
XM_017005170.1 | 199 | Missense Mutation | CAC,CAG | H,Q 29 | XP_016860659.1 |