Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTTTTTTTCAGCTGGTCCA[C/T]GTTGCTGGTCCTGGCCCCACAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607172 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK11IP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STK11IP - serine/threonine kinase 11 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052902.3 | 355 | Silent Mutation | CAC,CAT | H,H 92 | NP_443134.3 | |
XM_011510526.1 | 355 | Silent Mutation | CAC,CAT | H,H 92 | XP_011508828.1 | |
XM_011510527.2 | 355 | Silent Mutation | CAC,CAT | H,H 92 | XP_011508829.1 | |
XM_011510528.2 | 355 | Intron | XP_011508830.1 | |||
XM_011510529.1 | 355 | Silent Mutation | CAC,CAT | H,H 92 | XP_011508831.1 |