Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACCTGCTGAAGTGCAGCTTTGA[C/T]GTGGTCAAGCGCTGGGTGAATGAGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615525 | |||||||||||||||||||||||
Literature Links: |
COPG1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
COPG1 - coatomer protein complex subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016128.3 | 616 | Silent Mutation | GAC,GAT | D,D 172 | NP_057212.1 | |
XM_011512549.2 | 616 | Silent Mutation | GAC,GAT | D,D 172 | XP_011510851.1 |