Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTCCTTGCTGGAGTTTATTCC[A/G]ATTCCCTTTTGTCTGATTCTTCCTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609010 | |||||||||||||||||||||||
Literature Links: |
MCCC1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MCCC1 - methylcrotonoyl-CoA carboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293273.1 | 2124 | Missense Mutation | TCG,TTG | S,L 607 | NP_001280202.1 | |
NM_020166.4 | 2124 | Missense Mutation | TCG,TTG | S,L 724 | NP_064551.3 | |
XM_006713702.1 | 2124 | Missense Mutation | TCG,TTG | S,L 615 | XP_006713765.1 | |
XM_011512992.2 | 2124 | Missense Mutation | TCG,TTG | S,L 686 | XP_011511294.1 | |
XM_011512993.1 | 2124 | Intron | XP_011511295.1 | |||
XM_017006874.1 | 2124 | Missense Mutation | TCG,TTG | S,L 615 | XP_016862363.1 | |
XM_017006875.1 | 2124 | Missense Mutation | TCG,TTG | S,L 615 | XP_016862364.1 |