Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATGTCTGGCTGCTTCCCATTAC[A/G]GATTACTCTCAGCACATTGGAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605128 | ||||||||||||||||||||
Literature Links: |
GUCA1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GUCA1C - guanylate cyclase activator 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005459.3 | 615 | Missense Mutation | CGT,TGT | R,C 183 | NP_005450.3 | |
XM_005247910.3 | 615 | Missense Mutation | CCG,CTG | P,L 196 | XP_005247967.1 | |
XM_011513334.2 | 615 | Missense Mutation | CGT,TGT | R,C 99 | XP_011511636.1 |