Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCAGAGATGTTGTCCTCAGTTA[A/T]GCTCCTGTCTGTGCTGCAAGTTCAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TRIM42 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TRIM42 - tripartite motif containing 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152616.4 | 270 | Missense Mutation | AGC,TGC | S,C 22 | NP_689829.3 | |
XM_011512740.2 | 270 | Missense Mutation | AGC,TGC | S,C 22 | XP_011511042.1 |