Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGAGGCATCAAGAAGTCCGGCA[C/T]GGCAACTGAGAACCTATGGCTTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610122 | ||||||||||||||||||||
Literature Links: |
HTR3D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HTR3D - 5-hydroxytryptamine receptor 3D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145143.1 | 335 | Missense Mutation | ACG,ATG | T,M 51 | NP_001138615.1 | |
NM_001163646.1 | 335 | Missense Mutation | ACG,ATG | T,M 112 | NP_001157118.1 | |
NM_182537.2 | 335 | Intron | NP_872343.2 | |||
XM_017005854.1 | 335 | Intron | XP_016861343.1 |