Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTGCCTTGCTTCGTTGGGCCTA[A/T]GCAAGAACAATAAATGTCTATCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603840 | ||||||||||||||||||||
Literature Links: |
NDUFB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFB4 - NADH:ubiquinone oxidoreductase subunit B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168331.1 | 289 | Nonsense Mutation | TAA,TAT | *,Y 70 | NP_001161803.1 | |
NM_004547.5 | 289 | Nonsense Mutation | TAA,TAT | *,Y 70 | NP_004538.2 |