Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAGGATCAATGGTTGTTTCCC[A/G]TTGAATTTGTTCCATTGTTTCTGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615586 MIM: 610276 | |||||||||||||||||||||||
Literature Links: |
CEP19 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CEP19 - centrosomal protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032898.4 | 455 | Missense Mutation | NP_116287.2 | |||
XM_005269370.4 | 455 | Missense Mutation | XP_005269427.1 | |||
XM_011513246.2 | 455 | Missense Mutation | XP_011511548.1 |
PIGX - phosphatidylinositol glycan anchor biosynthesis class X | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |