Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTCCTAGTTAAAACTTCATGG[C/T]GCTTTGTTTCATGAATCTGGTCCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
RABL3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RABL3 - RAB, member of RAS oncogene family like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173825.3 | 484 | Missense Mutation | CAC,CGC | H,R 157 | NP_776186.2 | |
XM_006713595.2 | 484 | Missense Mutation | CAC,CGC | H,R 157 | XP_006713658.1 | |
XM_017006227.1 | 484 | Missense Mutation | CAC,CGC | H,R 157 | XP_016861716.1 | |
XM_017006228.1 | 484 | Missense Mutation | CAC,CGC | H,R 157 | XP_016861717.1 |