Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGCGCTGCCCTGCCCTGCACC[C/T]ACCCTCACTCTCCGTGTTCTCACTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609137 | |||||||||||||||||||||||
Literature Links: |
LOC101929106 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LOC101929106 - uncharacterized LOC101929106 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTP1 - receptor transporter protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153708.2 | 77 | Missense Mutation | CCA,CTA | P,L 16 | NP_714919.2 |