Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGCAGTGGCAGGCAGTCCACGT[C/T]GGCCACCAGACGGTTGATGACAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602441 MIM: 602130 | ||||||||||||||||||||
Literature Links: |
CISH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CISH - cytokine inducible SH2 containing protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013324.5 | 3712 | Missense Mutation | AAC,GAC | N,D 254 | NP_037456.5 | |
NM_145071.2 | 3712 | Missense Mutation | AAC,GAC | N,D 237 | NP_659508.1 | |
XM_011533329.2 | 3712 | Missense Mutation | AAC,GAC | N,D 243 | XP_011531631.1 |
MAPKAPK3 - mitogen-activated protein kinase-activated protein kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |