Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTCTGAGGTTTTGGTGGCCATT[A/C]TGGATCAACCAAGAGCTCCTGCGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610232 MIM: 173511 | ||||||||||||||||||||
Literature Links: |
ATP13A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP13A3 - ATPase 13A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024524.3 | 4124 | Nonsense Mutation | GAA,TAA | E,* 1199 | NP_078800.3 | |
XM_005269357.3 | 4124 | Nonsense Mutation | GAA,TAA | E,* 1229 | XP_005269414.1 | |
XM_011513120.2 | 4124 | Nonsense Mutation | GAA,TAA | E,* 1229 | XP_011511422.1 | |
XM_011513121.1 | 4124 | Nonsense Mutation | GAA,TAA | E,* 1229 | XP_011511423.1 | |
XM_011513122.1 | 4124 | Nonsense Mutation | GAA,TAA | E,* 1202 | XP_011511424.1 | |
XM_011513123.2 | 4124 | Nonsense Mutation | GAA,TAA | E,* 1199 | XP_011511425.1 | |
XM_011513124.2 | 4124 | Intron | XP_011511426.1 |
GP5 - glycoprotein V platelet | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |