Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTACGAAGGGCTCTGGAGGAGCTG[C/T]GTGAGGCAGAGTTCAGGCTTCACCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609210 | |||||||||||||||||||||||
Literature Links: |
CLDN18 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CLDN18 - claudin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002026.2 | 221 | Intron | NP_001002026.1 | |||
NM_016369.3 | 221 | Silent Mutation | TGC,TGT | C,C 53 | NP_057453.1 |