Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCTCTAAGGATGGGCTCTTCC[A/G]AGTGGACAAGGGTGCAGGCAACAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604543 | ||||||||||||||||||||
Literature Links: |
LIMD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIMD1 - LIM domains containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014240.2 | 138 | Missense Mutation | CAA,CGA | Q,R 30 | NP_055055.1 | |
XM_011534207.2 | 138 | Missense Mutation | CAA,CGA | Q,R 30 | XP_011532509.1 |