Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCCCCTGAGCCAGGGCGCGGGG[A/G]TTAAAGGTGCCGTTGGGGTCTCGCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602142 | |||||||||||||||||||||||
Literature Links: |
PLCD1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLCD1 - phospholipase C delta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130964.1 | 2235 | Silent Mutation | AAC,AAT | N,N 637 | NP_001124436.1 | |
NM_006225.3 | 2235 | Silent Mutation | AAC,AAT | N,N 616 | NP_006216.2 | |
XM_017006622.1 | 2235 | Silent Mutation | AAC,AAT | N,N 558 | XP_016862111.1 |
VILL - villin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |