Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGGTCTTTCTCCCTTGCATAC[A/G]TGGTAGCTTTTCTGTGAATTTTTTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611318 MIM: 600515 MIM: 606380 | |||||||||||||||||||||||
Literature Links: |
MED12L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MED12L - mediator complex subunit 12 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053002.5 | 1009 | Intron | NP_443728.3 | |||
XM_006713487.3 | 1009 | Intron | XP_006713550.1 | |||
XM_011512390.2 | 1009 | Intron | XP_011510692.1 | |||
XM_011512394.2 | 1009 | Intron | XP_011510696.1 | |||
XM_011512399.2 | 1009 | Intron | XP_011510701.1 | |||
XM_017005676.1 | 1009 | Intron | XP_016861165.1 | |||
XM_017005677.1 | 1009 | Intron | XP_016861166.1 | |||
XM_017005678.1 | 1009 | Intron | XP_016861167.1 | |||
XM_017005679.1 | 1009 | Intron | XP_016861168.1 | |||
XM_017005680.1 | 1009 | Intron | XP_016861169.1 | |||
XM_017005681.1 | 1009 | Intron | XP_016861170.1 |
P2RY12 - purinergic receptor P2Y12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P2RY13 - purinergic receptor P2Y13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176894.2 | 1009 | Missense Mutation | CGT,TGT | R,C 330 | NP_795713.2 | |
XM_006713664.1 | 1009 | Intron | XP_006713727.1 |