Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTCCCATGCCTGCCTGGGGAGC[C/T]CTGTTCCTGCTCTGGGCCACAGCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 173515 | |||||||||||||||||||||||
Literature Links: |
GP9 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GP9 - glycoprotein IX platelet | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000174.4 | 493 | Silent Mutation | GCC,GCT | A,A 6 | NP_000165.1 | |
XM_005247374.3 | 493 | Silent Mutation | GCC,GCT | A,A 6 | XP_005247431.1 | |
XM_011512701.1 | 493 | Silent Mutation | GCC,GCT | A,A 6 | XP_011511003.1 | |
XM_011512702.1 | 493 | Silent Mutation | GCC,GCT | A,A 6 | XP_011511004.1 |