Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAAAGGAAAACCACTAGCATTTC[A/G]AGTAGGTCAGTCTTTTTTGTGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMAA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMAA - methylmalonic aciduria (cobalamin deficiency) cblA type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172250.2 | 597 | Missense Mutation | CAA,CGA | Q,R 145 | NP_758454.1 | |
XM_011531684.2 | 597 | Missense Mutation | CAA,CGA | Q,R 145 | XP_011529986.1 | |
XM_011531685.2 | 597 | Missense Mutation | CAA,CGA | Q,R 145 | XP_011529987.1 | |
XM_011531686.2 | 597 | UTR 5 | XP_011529988.1 |