Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTAAGCTAGAGTCCACGGACT[A/C]TTTATCACACTTTTCACTGAGCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606645 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARAP2 - ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015230.3 | 5428 | Nonsense Mutation | GAG,TAG | E,* 1599 | NP_056045.2 | |
XM_017007698.1 | 5428 | Intron | XP_016863187.1 | |||
XM_017007699.1 | 5428 | Nonsense Mutation | GAG,TAG | E,* 1599 | XP_016863188.1 | |
XM_017007700.1 | 5428 | Nonsense Mutation | GAG,TAG | E,* 1576 | XP_016863189.1 | |
XM_017007701.1 | 5428 | Nonsense Mutation | GAG,TAG | E,* 1554 | XP_016863190.1 | |
XM_017007702.1 | 5428 | Nonsense Mutation | GAG,TAG | E,* 1531 | XP_016863191.1 |