Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATAAACTTGAAAATGCAAGATT[A/G]AGTAGAGAAAATAATGAATTATACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP135 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP135 - centrosomal protein 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025009.4 | 502 | Silent Mutation | TTA,TTG | L,L 78 | NP_079285.2 | |
XM_005265788.3 | 502 | UTR 5 | XP_005265845.1 | |||
XM_006714055.2 | 502 | Silent Mutation | TTA,TTG | L,L 78 | XP_006714118.1 | |
XM_011534412.2 | 502 | Intron | XP_011532714.1 |