Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATCTCATAGCGGTAATGCCGGAA[C/T]TCCACGGCAAAGGTGCCAAAATAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC105377310 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105377310 - syncytin-1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS6P6 - ribosomal protein S6 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM150C - transmembrane protein 150C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080506.1 | 720 | Silent Mutation | GAA,GAG | E,E 214 | NP_001073975.1 |