Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTTCTTAATCTTAATAATGGTC[A/C]ACTTTTGCCACTACAACTTCAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614843 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ODAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ODAM - odontogenic, ameloblast asssociated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017855.3 | 136 | Missense Mutation | CAA,CCA | Q,P 40 | NP_060325.3 | |
XM_005265698.3 | 136 | Missense Mutation | CAA,CCA | Q,P 57 | XP_005265755.1 | |
XM_006714245.3 | 136 | Missense Mutation | CAA,CCA | Q,P 57 | XP_006714308.1 | |
XM_017008331.1 | 136 | Missense Mutation | CAA,CCA | Q,P 26 | XP_016863820.1 |