Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAGTTCATACAACTGAGAGCCTT[C/T]GGCAATCAAAATTATCTGCTGTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611423 | ||||||||||||||||||||
Literature Links: |
CEP135 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP135 - centrosomal protein 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025009.4 | 410 | Missense Mutation | CGG,TGG | R,W 48 | NP_079285.2 | |
XM_005265788.3 | 410 | UTR 5 | XP_005265845.1 | |||
XM_006714055.2 | 410 | Missense Mutation | CGG,TGG | R,W 48 | XP_006714118.1 | |
XM_011534412.2 | 410 | Intron | XP_011532714.1 |