Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGTGAAGAATGTGGCAGCATCT[G/T]TACCACATCCTCACACTTTGCTAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194648 | ||||||||||||||||||||
Literature Links: |
ZNF141 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF141 - zinc finger protein 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003441.2 | 1473 | Missense Mutation | TGT,TTT | C,F 236 | NP_003432.1 | |
XM_011513562.2 | 1473 | Missense Mutation | TGT,TTT | C,F 264 | XP_011511864.1 | |
XM_011513563.2 | 1473 | Missense Mutation | TGT,TTT | C,F 160 | XP_011511865.1 | |
XM_017008591.1 | 1473 | Missense Mutation | TGT,TTT | C,F 296 | XP_016864080.1 |