Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACATTCCAGAATTCCCTTATTC[A/G]TCTTGGACTCAACACTATGAAGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606261 MIM: 613940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT6 - nudix hydrolase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA5 - spermatogenesis associated 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317799.1 | 276 | Missense Mutation | CAT,CGT | H,R 68 | NP_001304728.1 | |
NM_145207.2 | 276 | Missense Mutation | CAT,CGT | H,R 69 | NP_660208.2 | |
XM_005262783.4 | 276 | Missense Mutation | CAT,CGT | H,R 68 | XP_005262840.1 | |
XM_011531678.2 | 276 | Missense Mutation | CAT,CGT | H,R 68 | XP_011529980.1 | |
XM_011531679.2 | 276 | Missense Mutation | CAT,CGT | H,R 69 | XP_011529981.1 | |
XM_017007825.1 | 276 | Missense Mutation | CAT,CGT | H,R 69 | XP_016863314.1 | |
XM_017007826.1 | 276 | Missense Mutation | CAT,CGT | H,R 69 | XP_016863315.1 | |
XM_017007827.1 | 276 | Missense Mutation | CAT,CGT | H,R 69 | XP_016863316.1 | |
XM_017007828.1 | 276 | UTR 5 | XP_016863317.1 | |||
XM_017007829.1 | 276 | Intron | XP_016863318.1 | |||
XM_017007830.1 | 276 | Missense Mutation | CAT,CGT | H,R 69 | XP_016863319.1 |