Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACCTGCGTTTTGATCTGGATAA[A/G]CATATTGTCCATCTCCCACAGCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605695 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLOC1S4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BLOC1S4 - biogenesis of lysosomal organelles complex 1 subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRFAP1L1 - Morf4 family associated protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203462.2 | 440 | Missense Mutation | CTT,TTT | L,F 64 | NP_982287.1 | |
XM_005247932.1 | 440 | Missense Mutation | CTT,TTT | L,F 64 | XP_005247989.1 |