Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGGCAGAATATTTTCATATTCT[A/G]GACGTGCACGGCAAGAACACCTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603919 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB9 - EF-hand calcium binding domain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171183.1 | 109 | Silent Mutation | CTA,CTG | L,L 36 | NP_001164654.1 | |
XM_017009350.1 | 109 | Intron | XP_016864839.1 |
STK10 - serine/threonine kinase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |