Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATATATATAGGTTATTTGTAGC[C/T]GCCACTGCTATAATATTTTCTGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP2R2B - protein phosphatase 2 regulatory subunit Bbeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271899.1 | 1887 | Silent Mutation | GCA,GCG | A,A 436 | NP_001258828.1 | |
NM_001271900.1 | 1887 | Silent Mutation | GCA,GCG | A,A 488 | NP_001258829.1 | |
NM_001271948.1 | 1887 | Silent Mutation | GCA,GCG | A,A 419 | NP_001258877.1 | |
NM_181674.2 | 1887 | Silent Mutation | GCA,GCG | A,A 496 | NP_858060.2 | |
NM_181675.3 | 1887 | Silent Mutation | GCA,GCG | A,A 536 | NP_858061.2 | |
NM_181676.2 | 1887 | Silent Mutation | GCA,GCG | A,A 433 | NP_858062.1 | |
NM_181677.2 | 1887 | Silent Mutation | GCA,GCG | A,A 410 | NP_858063.1 | |
NM_181678.2 | 1887 | Silent Mutation | GCA,GCG | A,A 419 | NP_858064.1 |