Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATTTCATTCATTGTCTGTATTA[C/T]GAAGCCCACATCCCAGTTCAAAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606694 | ||||||||||||||||||||
Literature Links: |
NUP155 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUP155 - nucleoporin 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278312.1 | 4002 | Missense Mutation | ATA,GTA | I,V 1221 | NP_001265241.1 | |
NM_004298.3 | 4002 | Missense Mutation | ATA,GTA | I,V 1226 | NP_004289.1 | |
NM_153485.2 | 4002 | Missense Mutation | ATA,GTA | I,V 1285 | NP_705618.1 | |
XM_011514165.2 | 4002 | Intron | XP_011512467.1 |