Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAATGAAGATGTGATTATTAAC[G/T]GTCATTCTCATGAAGATGACAATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605604 MIM: 603790 | ||||||||||||||||||||
Literature Links: |
PAIP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAIP2 - poly(A) binding protein interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033112.2 | 1647 | Missense Mutation | GGT,TGT | G,C 22 | NP_001028284.1 | |
NM_016480.4 | 1647 | Missense Mutation | GGT,TGT | G,C 22 | NP_057564.3 | |
XM_011543428.1 | 1647 | Missense Mutation | GGT,TGT | G,C 22 | XP_011541730.1 | |
XM_017009522.1 | 1647 | Missense Mutation | GGT,TGT | G,C 22 | XP_016865011.1 | |
XM_017009523.1 | 1647 | Missense Mutation | GGT,TGT | G,C 22 | XP_016865012.1 |
SLC23A1 - solute carrier family 23 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |