Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGTAGCATTAAAGCTCTATCC[C/T]GTCTTGCCGATCTTGGTTATGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf24 - chromosome 5 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135586.1 | 864 | Missense Mutation | CGT,TGT | R,C 150 | NP_001129058.1 | |
NM_001300894.1 | 864 | Intron | NP_001287823.1 | |||
NM_152409.3 | 864 | Missense Mutation | CGT,TGT | R,C 150 | NP_689622.2 | |
XM_005271889.3 | 864 | Missense Mutation | CGT,TGT | R,C 150 | XP_005271946.1 | |
XM_017009049.1 | 864 | Missense Mutation | CGT,TGT | R,C 150 | XP_016864538.1 | |
XM_017009050.1 | 864 | Missense Mutation | CGT,TGT | R,C 150 | XP_016864539.1 |