Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCCTGCCCCACAGCTTCTAAG[C/T]GCCAGATGATGGAGGAGCGTGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUFY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RUFY1 - RUN and FYVE domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040451.2 | 328 | UTR 5 | NP_001035541.1 | |||
NM_001040452.2 | 328 | UTR 5 | NP_001035542.1 | |||
NM_025158.4 | 328 | Missense Mutation | CGC,TGC | R,C 107 | NP_079434.3 | |
XM_005265993.4 | 328 | Intron | XP_005266050.1 | |||
XM_006714921.3 | 328 | Missense Mutation | CGC,TGC | R,C 107 | XP_006714984.2 | |
XM_006714922.3 | 328 | Intron | XP_006714985.1 | |||
XM_017009891.1 | 328 | UTR 5 | XP_016865380.1 | |||
XM_017009892.1 | 328 | Intron | XP_016865381.1 | |||
XM_017009893.1 | 328 | Intron | XP_016865382.1 | |||
XM_017009894.1 | 328 | UTR 5 | XP_016865383.1 | |||
XM_017009895.1 | 328 | UTR 5 | XP_016865384.1 |