Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCATCTGCATGATGTCCATGAAC[A/G]TGTTTACTGGTAACAATAAGATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142910 | ||||||||||||||||||||
Literature Links: |
HMGCR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMGCR - 3-hydroxy-3-methylglutaryl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000859.2 | 262 | Missense Mutation | ATG,GTG | M,V 36 | NP_000850.1 | |
NM_001130996.1 | 262 | Missense Mutation | ATG,GTG | M,V 36 | NP_001124468.1 | |
XM_011543357.1 | 262 | Missense Mutation | ATG,GTG | M,V 56 | XP_011541659.1 | |
XM_011543358.1 | 262 | Missense Mutation | ATG,GTG | M,V 36 | XP_011541660.1 | |
XM_011543359.1 | 262 | Missense Mutation | ATG,GTG | M,V 56 | XP_011541661.1 |