Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAGCGCCGCTCCAGCTCATAGA[C/T]CTGCGCCTGCGAGAAGAGCACGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKX2-5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NKX2-5 - NK2 homeobox 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166175.1 | 932 | UTR 3 | NP_001159647.1 | |||
NM_001166176.1 | 932 | UTR 3 | NP_001159648.1 | |||
NM_004387.3 | 932 | Missense Mutation | ATC,GTC | I,V 150 | NP_004378.1 | |
XM_017009071.1 | 932 | Intron | XP_016864560.1 |