Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGTAGCCGGCCAGCCAGCCCTC[A/G]TAACCTAGCACCAGAGCACCCCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VDAC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VDAC1 - voltage dependent anion channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003374.2 | 683 | Silent Mutation | TAC,TAT | Y,Y 146 | NP_003365.1 | |
XM_005272075.3 | 683 | Intron | XP_005272132.1 | |||
XM_017009821.1 | 683 | Intron | XP_016865310.1 | |||
XM_017009822.1 | 683 | Intron | XP_016865311.1 | |||
XM_017009823.1 | 683 | Intron | XP_016865312.1 |