Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGCCCATGCCCCCACCAGGGCC[C/T]TCGTGAGGACGATGCCCACCACCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603771 | ||||||||||||||||||||
Literature Links: |
PPP1R10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R10 - protein phosphatase 1 regulatory subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002714.3 | 2871 | Silent Mutation | GAA,GAG | E,E 755 | NP_002705.2 | |
XM_006715130.2 | 2871 | Silent Mutation | GAA,GAG | E,E 755 | XP_006715193.1 | |
XM_011514722.1 | 2871 | Silent Mutation | GAA,GAG | E,E 755 | XP_011513024.1 | |
XM_017010995.1 | 2871 | Silent Mutation | GAA,GAG | E,E 755 | XP_016866484.1 |