Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGTTTTCTCTTGGATCAAGGC[A/G]AGCTTGGCATCTTGTTCCTCAGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138360 | ||||||||||||||||||||
Literature Links: |
GSTA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTA2 - glutathione S-transferase alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000846.4 | 513 | Silent Mutation | CTC,CTT | L,L 121 | NP_000837.3 | |
XM_011514532.2 | 513 | Intron | XP_011512834.1 |
GSTA7P - glutathione S-transferase alpha 7, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |