Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTACCCAAACACACACTGGGGAG[C/G]TCGGGAAGGTATCTGCAGGTCGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606915 | ||||||||||||||||||||
Literature Links: |
GPR63 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR63 - G protein-coupled receptor 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143957.2 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | NP_001137429.1 | |
NM_030784.3 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | NP_110411.1 | |
XM_006715570.3 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | XP_006715633.1 | |
XM_011536153.2 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | XP_011534455.1 | |
XM_011536154.2 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | XP_011534456.1 | |
XM_011536155.2 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | XP_011534457.1 | |
XM_011536157.2 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | XP_011534459.1 | |
XM_011536159.2 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | XP_011534461.1 | |
XM_017011334.1 | 1564 | Missense Mutation | CCT,GCT | P,A 226 | XP_016866823.1 |