Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTGCTTGGAAACAGGTGAGCTG[C/T]AAGTGCATGCTACTCTTGAACTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615329 | ||||||||||||||||||||
Literature Links: |
EXOC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXOC2 - exocyst complex component 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018303.5 | 2939 | Silent Mutation | TTA,TTG | L,L 909 | NP_060773.3 | |
XM_017011018.1 | 2939 | Silent Mutation | TTA,TTG | L,L 909 | XP_016866507.1 | |
XM_017011019.1 | 2939 | Silent Mutation | TTA,TTG | L,L 909 | XP_016866508.1 | |
XM_017011020.1 | 2939 | Silent Mutation | TTA,TTG | L,L 909 | XP_016866509.1 | |
XM_017011021.1 | 2939 | Silent Mutation | TTA,TTG | L,L 909 | XP_016866510.1 | |
XM_017011022.1 | 2939 | Silent Mutation | TTA,TTG | L,L 909 | XP_016866511.1 | |
XM_017011023.1 | 2939 | Silent Mutation | TTA,TTG | L,L 909 | XP_016866512.1 | |
XM_017011024.1 | 2939 | Silent Mutation | TTA,TTG | L,L 909 | XP_016866513.1 | |
XM_017011025.1 | 2939 | Intron | XP_016866514.1 | |||
XM_017011026.1 | 2939 | Intron | XP_016866515.1 |