Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGCATCTTTTGATCATTCTCC[A/G]GATAGCCTTCCTTTGAGGAGCTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613081 MIM: 187680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM1B - lysine demethylase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153042.3 | 295 | Silent Mutation | CCA,CCG | P,P 18 | NP_694587.3 | |
XM_005248925.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_005248982.1 | |
XM_005248926.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_005248983.1 | |
XM_005248928.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_005248985.1 | |
XM_011514385.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_011512687.1 | |
XM_011514386.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_011512688.1 | |
XM_017010440.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_016865929.1 | |
XM_017010441.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_016865930.1 | |
XM_017010442.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_016865931.1 | |
XM_017010443.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_016865932.1 | |
XM_017010445.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_016865934.1 | |
XM_017010446.1 | 295 | Silent Mutation | CCA,CCG | P,P 18 | XP_016865935.1 |
TPMT - thiopurine S-methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |