Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAAGGACCAGTTGATTTGTGAA[A/G]ATCAACAATATGACACCTATTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZUFSP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZUFSP - zinc finger with UFM1 specific peptidase domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145062.2 | 1986 | Missense Mutation | CTT,TTT | L,F 445 | NP_659499.2 | |
XM_011535565.2 | 1986 | Missense Mutation | CTT,TTT | L,F 445 | XP_011533867.1 | |
XM_011535567.1 | 1986 | Missense Mutation | CTT,TTT | L,F 277 | XP_011533869.1 | |
XM_017010393.1 | 1986 | Missense Mutation | CTT,TTT | L,F 445 | XP_016865882.1 | |
XM_017010394.1 | 1986 | Missense Mutation | CTT,TTT | L,F 249 | XP_016865883.1 | |
XM_017010395.1 | 1986 | Missense Mutation | CTT,TTT | L,F 221 | XP_016865884.1 | |
XM_017010396.1 | 1986 | Missense Mutation | CTT,TTT | L,F 221 | XP_016865885.1 |