Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGATGAGCCCCGGCACCCCCGGA[C/T]CGACCATGGGCAGATCCCAGGTAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614556 | ||||||||||||||||||||
Literature Links: |
ARID1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARID1B - AT-rich interaction domain 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017519.2 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | NP_059989.2 | |
NM_020732.3 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | NP_065783.3 | |
XM_005267069.4 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | XP_005267126.2 | |
XM_011535984.2 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | XP_011534286.2 | |
XM_011535988.2 | 1597 | Intron | XP_011534290.1 | |||
XM_017011103.1 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | XP_016866592.1 | |
XM_017011104.1 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | XP_016866593.1 | |
XM_017011105.1 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | XP_016866594.1 | |
XM_017011106.1 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | XP_016866595.1 | |
XM_017011107.1 | 1597 | Missense Mutation | CCG,TCG | P,S 508 | XP_016866596.1 | |
XM_017011108.1 | 1597 | Missense Mutation | CCG,TCG | P,S 535 | XP_016866597.1 | |
XM_017011109.1 | 1597 | Intron | XP_016866598.1 |
MIR4466 - microRNA 4466 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |