Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGGAATTATTACCTGGTCATT[A/C]TCTAACTTGGAATGCAGCTGGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605433 | ||||||||||||||||||||
Literature Links: |
KIF13A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF13A - kinesin family member 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105566.2 | 5519 | Missense Mutation | GAG,GAT | E,D 1753 | NP_001099036.1 | |
NM_001105567.2 | 5519 | Missense Mutation | GAG,GAT | E,D 1740 | NP_001099037.1 | |
NM_001105568.2 | 5519 | Missense Mutation | GAG,GAT | E,D 1740 | NP_001099038.1 | |
NM_001243423.1 | 5519 | Intron | NP_001230352.1 | |||
NM_022113.5 | 5519 | Missense Mutation | GAG,GAT | E,D 1788 | NP_071396.4 | |
XM_011514806.2 | 5519 | Intron | XP_011513108.1 | |||
XM_017011188.1 | 5519 | Missense Mutation | GAG,GAT | E,D 1788 | XP_016866677.1 | |
XM_017011189.1 | 5519 | Missense Mutation | GAG,GAT | E,D 1775 | XP_016866678.1 | |
XM_017011190.1 | 5519 | Missense Mutation | GAG,GAT | E,D 1767 | XP_016866679.1 | |
XM_017011191.1 | 5519 | Missense Mutation | GAG,GAT | E,D 1754 | XP_016866680.1 | |
XM_017011192.1 | 5519 | Missense Mutation | GAG,GAT | E,D 1753 | XP_016866681.1 | |
XM_017011193.1 | 5519 | Missense Mutation | GAG,GAT | E,D 1677 | XP_016866682.1 |