Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGCGCTGCGTGGGCCATGGTGCA[G/T]CTCCGACCGCGAGCGTCTCGCGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611533 MIM: 603854 MIM: 604483 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOL7 - nucleolar protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317724.1 | 41 | Silent Mutation | CAG,CAT | Q,H 3 | NP_001304653.1 | |
NM_016167.4 | 41 | Silent Mutation | CAG,CAT | Q,H 3 | NP_057251.2 |
RANBP9 - RAN binding protein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRT5 - sirtuin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193267.2 | 41 | Intron | NP_001180196.1 | |||
NM_001242827.1 | 41 | Intron | NP_001229756.1 | |||
NM_012241.4 | 41 | Intron | NP_036373.1 | |||
NM_031244.3 | 41 | Intron | NP_112534.1 | |||
XM_005248968.4 | 41 | Intron | XP_005249025.1 | |||
XM_011514444.2 | 41 | Intron | XP_011512746.1 | |||
XM_017010620.1 | 41 | Intron | XP_016866109.1 | |||
XM_017010621.1 | 41 | Intron | XP_016866110.1 | |||
XM_017010622.1 | 41 | Intron | XP_016866111.1 | |||
XM_017010623.1 | 41 | Intron | XP_016866112.1 | |||
XM_017010624.1 | 41 | Intron | XP_016866113.1 | |||
XM_017010625.1 | 41 | Intron | XP_016866114.1 |