Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACGGAACTATTTACTAAAGAGA[C/T]GCATAAAAAATCACCTGCAAGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612424 MIM: 607789 | ||||||||||||||||||||
Literature Links: |
EYS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EYS - eyes shut homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142800.1 | 8849 | Missense Mutation | ATC,GTC | I,V 2750 | NP_001136272.1 | |
NM_001142801.1 | 8849 | Intron | NP_001136273.1 | |||
NM_001292009.1 | 8849 | Missense Mutation | ATC,GTC | I,V 2771 | NP_001278938.1 | |
NM_198283.1 | 8849 | Intron | NP_938024.1 |
PHF3 - PHD finger protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |