Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCTTCACAGTTACTTGGGCAGG[C/T]TTCACATGGGACGCCTGTCTTATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601193 | ||||||||||||||||||||
Literature Links: |
CRISP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRISP1 - cysteine rich secretory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131.2 | 617 | Missense Mutation | ACC,GCC | T,A 197 | NP_001122.2 | |
NM_001205220.1 | 617 | Missense Mutation | ACC,GCC | T,A 197 | NP_001192149.1 | |
NM_170609.1 | 617 | Intron | NP_733758.1 | |||
XM_011514315.2 | 617 | Missense Mutation | ACC,GCC | T,A 95 | XP_011512617.1 | |
XM_017010320.1 | 617 | Missense Mutation | ACC,GCC | T,A 95 | XP_016865809.1 | |
XM_017010321.1 | 617 | Missense Mutation | ACC,GCC | T,A 95 | XP_016865810.1 |