Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTAAAGAAAAAGAAGAAATACA[A/G]TACCATCTCAACTTCCTGAAGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611506 MIM: 609058 | ||||||||||||||||||||
Literature Links: |
CENPQ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPQ - centromere protein Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018132.3 | 346 | Silent Mutation | CAA,CAG | Q,Q 107 | NP_060602.2 | |
XM_005249205.1 | 346 | Silent Mutation | CAA,CAG | Q,Q 107 | XP_005249262.1 | |
XM_011514723.2 | 346 | Silent Mutation | CAA,CAG | Q,Q 110 | XP_011513025.2 |
MUT - methylmalonyl-CoA mutase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |