Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGTGCTCCGTGTAGGTAACTGC[A/G]TCGCGGATTACATTCTCCAAAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142711 MIM: 602793 MIM: 602814 MIM: 602831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST1H1B - histone cluster 1, H1b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2AL - histone cluster 1, H2al | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H3I - histone cluster 1, H3i | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003533.2 | 208 | Intron | NP_003524.1 |
HIST1H4L - histone cluster 1, H4l | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003546.2 | 208 | Silent Mutation | GAC,GAT | D,D 69 | NP_003537.1 |