Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCACTCAGGCCCAGGTGGACCA[C/T]TATTTTGTCTTGCCTTTCTTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137162 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRR2 - gamma-aminobutyric acid type A receptor rho2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002043.4 | 2441 | Missense Mutation | ATG,GTG | M,V 408 | NP_002034.3 | |
XM_011535713.2 | 2441 | Missense Mutation | ATG,GTG | M,V 364 | XP_011534015.1 | |
XM_011535714.2 | 2441 | Missense Mutation | ATG,GTG | M,V 309 | XP_011534016.1 | |
XM_011535715.2 | 2441 | Missense Mutation | ATG,GTG | M,V 309 | XP_011534017.1 | |
XM_011535716.2 | 2441 | Missense Mutation | ATG,GTG | M,V 309 | XP_011534018.1 | |
XM_011535717.2 | 2441 | Missense Mutation | ATG,GTG | M,V 309 | XP_011534019.1 |